Description
- Installation and operational qualification documents for validation performed by customers
- Designed for use with APT-COM™ GLP-Edition software
- Includes IQ/OQ checklists, calibration guide, and comprehensive unit documentation
- Both digital and hard copy options are available
Specifications
Specifications
| For Use With (Equipment) | Multi Management Software APT-COM™ GLP-Edition Software |
| Description | Qualification documents - Digital in PDF format, IQ/OQ documents - supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; for APT-COM GLP software |
| Product Type | IQ/OQ Documentation (PDF) |
| Product Line | APT-COM |
| Service Offering | GxP Compliant IQ/OQ Documentation |
| For Use With (Application) | Installation and Operational Qualification |
| Includes | IQ/OQ Checklists with Calibration Guide and Comprehensive Unit Documentation |