Description
Our IQ/OQ Arium™ Comfort I H2O-I-2-TOC includes:
- On-site qualification (IQ OQ) by a specially trained service technician
- Performance of device-specific tests for the Installation Qualification (IQ)
- Checking the equipment delivery for completeness and any transport damage
- Documentation of installation conditions
- Verification of correct installation of equipment components
- Performance of device-specific tests for the Operational Qualification (OQ)
- Function tests
- Metrological equipment checks and calibration
- Training of the operating personnel
- Complete documentation in special IQ OQ templates
- Consultation on device maintenance and maintenance cycles
Specifications
Specifications
| For Use With (Equipment) | Arium Water Purification System |
| Product Type | IQ/OQ Service |