Learn More
Memmert™ Iq/Oq Document With Device-Specific Works Test Data (1 Temperature Value), For Ipp55
Product Code. 16313787 Shop All Memmert Products
Product Code. 16313787 Supplier Memmert™ Supplier No. IPP55D00127
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes temperature distribution survey for one free-selectable temperature value.
  • IQ/OQ document with works test data for chamber
  • 9 measuring points (size 30), 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand), PQ check list as support for validation by customer
Specifications
Product Type IQ/OQ Documentation