Learn More
Thermo Scientific IQ/OQ Document Pack for Advanced Centrifugal Evaporator
Product Code. 32018034 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Advanced Centrifugal Evaporator
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
32018034 Advanced Centrifugal Evaporator Each
1 options
Product Code. 32018034 Supplier Thermo Scientific Supplier No. IQOQDOCADVEVAPMVS
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides the IQ/OQ documentation pack for advanced centrifugal evaporators where required for qualification execution.
  • This SKU covers the document pack only and does not include on-site labor, travel, execution of qualification tests, installation, commissioning, preventative maintenance, spare parts or consumables
  • On-site IQ/OQ execution must be ordered separately using the applicable IQ/OQ execution SKU
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Advanced Centrifugal Evaporator
Product Type IQ/OQ Documentation