Learn More
Memmert™ Iq Document With Device-Specific Works Test Data For Validation By Customer, For U750
Shop All Memmert ProductsDescription
Includes
Works test data
- OQ/PQ check list as support for validation by customer
Specifications
Specifications
| For Use With (Equipment) | 30, 55, 75, 110, 160, 260, 450, 750, 1060 Models Climate Chamber |
| Product Type | IQ Document |