missing translation for 'onlineSavingsMsg'
Learn More

IQ 1, MedChemExpress

Product Code. 16492936
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492936 25 mg 25mg
16480417 50 mg 50mg
16436767 5 mg 5mg
16414068 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492936 Supplier Medchem Express Supplier No. HY10593
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress IQ 1 has many functions such as decreasing Wnt-stimulated phosphorylation, maintaining the pluripotency of murine ESCs, preventing PP2A/Nkd interaction and so on.

IQ 1 has many functions such as decreasing Wnt-stimulated phosphorylation, maintaining the pluripotency of murine ESCs, preventing PP2A/Nkd interaction and so on.

Specifications

Chemical Name or Material IQ 1
CAS 331001-62-8
Color Orange
Purity Grade Notes Research
Molecular Formula C21H22N4O2
Quantity 25 mg
Solubility Information DMSO : ≥ 33 mg/mL (91.05 mM)
SMILES CC(C1=CC=C(/N=N/C(C(N)=O)=C2NC(C)(C)CC3=CC=CC=C3/2)C=C1)=O
Molecular Weight (g/mol) 362.42
Formula Weight 362.42
Percent Purity 99.69%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.