missing translation for 'onlineSavingsMsg'
Learn More

Melford IPTG, Dioxane Free, Non-Mammalian

Product Code. 30018362
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30018362 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30018362

Brand: Melford I561005.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • Substance Name: Isopropyl-B-D-thio-galactopyranoside ex plant origin D-galactose
  • CAS Number: 367-93-1
  • Molecular Formula: C9H18O5S
  • Molecular Weight: 238.3

Specifications

CAS 367-93-1
Molecular Formula C9H18O5S
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.