missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress IPTG

Product Code. 16493506
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493506 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16493506 Supplier MedChemExpress Supplier No. HY15921
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress IPTG is a molecular mimic of allolactose, a lactose metabolite that triggers transcription of the lac operon, and it is therefore used to induce protein expression where the gene is under the control of the lac operator.

Specifications

CAS 367-93-1
Solubility H2O : 100 mg/mL (419.64 mM∣Need ultrasonic) ∣DMSO : ≥ 60 mg/mL (251.78 mM)
Synonym Isopropyl β-D-thiogalactoside
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Quantity 25 g
Molecular Weight (g/mol) 238.3 g/mol
Product Type Biochemical Assay Reagents
Purity or Quality Grade 95.0%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.