missing translation for 'onlineSavingsMsg'
Learn More

Melford IPTG

Product Code. 30018359
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
100 g
Unit Size:
100g
1g
25g
50g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018359 25 g 25g
30018361 50 g 50g
30018358 100 g 100g
30018360 5 g 5g
30018357 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30018359 Supplier Melford Supplier No. I5600025.0
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay.

IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay. Distinguishes recombinants (white) from non- recombinants (blue) in cloning strategies using vectors such as Lambda-11, M13mp 18 and 19, pUC18 and 19, pUR222 containing the lacZ gene.

Dioxane-Free

Water Content: <0.49%

Specifications

CAS 367-93-1
Molecular Formula C9H18O5S
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.