missing translation for 'onlineSavingsMsg'
Learn More

Iprovalicarb-d8, TRC

Product Code. 30397368
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397368 25 mg 25mg
30398662 10 mg 10mg
30391646 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30397368 Supplier Toronto Research Chemicals Supplier No. TRCI74750225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Iprovalicarb-d8
Molecular Formula C18 D8 H20 N2 O3
Quantity 25 mg
Synonym Iprovalicarb D8 (Valinyl D8)
InChI Formula InChI=1S/C18H28N2O3/c1-11(2)16(20-18(22)23-12(3)4)17(21)19-14(6)15-9-7-13(5)8-10-15/h7-12,14,16H,1-6H3,(H,19,21)(H,20,22)/t14?,16-/m0/s1/i1D3,2D3,11D,16D
SMILES [2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])[C@]([2H])(NC(=O)OC(C)C)C(=O)NC(C)c1ccc(C)cc1
IUPAC Name propan-2-yl N-[(2S)-2,3,4,4,4-pentadeuterio-1-[1-(4-methylphenyl)ethylamino]-1-oxo-3-(trideuteriomethyl)butan-2-yl]carbamate
Molecular Weight (g/mol) 328.4758
Formula Weight 328.2602
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.