missing translation for 'onlineSavingsMsg'
Learn More

Ipronidazole-d3, TRC

Product Code. 30138143
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138143 50 mg 50mg
17977649 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30138143 Supplier Toronto Research Chemicals Supplier No. TRCI74700250MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1015855-83-0
Molecular Formula C7 2H3 H8 N3 O2
Molecular Weight (g/mol) 172.2
Synonym 1H-Imidazole, 1-(methyl-d3)-2-(1-methylethyl)-5-nitro- (ACI), 1-(Methyl-d3)-2-(1-methylethyl)-5-nitro-1H-imidazole (ACI), Ipronidazole D3, Ipronidazole-D3 (N-methyl-D3), Ipronidazole-d3
IUPAC Name 5-nitro-2-propan-2-yl-1-(trideuteriomethyl)imidazole
SMILES [2H]C([2H])([2H])n1c(ncc1[N+](=O)[O-])C(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H11N3O2/c1-5(2)7-8-4-6(9(7)3)10(11)12/h4-5H,1-3H3/i3D3
Formula Weight 172.104
Chemical Name or Material Ipronidazole D3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.