missing translation for 'onlineSavingsMsg'
Learn More

Iprodione-d5, TRC

Product Code. 30402503
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402503 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30402503 Supplier Toronto Research Chemicals Supplier No. TRCI7450021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iprodione-d5
CAS 1215631-57-4
Molecular Formula C13 2H5 H8 Cl2 N3 O3
Quantity 1 mg
Synonym Iprodione D5 (dichlorophenyl D3 hydantoin D2)
InChI Formula InChI=1S/C13H13Cl2N3O3/c1-7(2)16-12(20)17-6-11(19)18(13(17)21)10-4-8(14)3-9(15)5-10/h3-5,7H,6H2,1-2H3,(H,16,20)/i3D,4D,5D,6D2
SMILES [2H]c1c(Cl)c([2H])c(N2C(=O)N(C(=O)NC(C)C)C([2H])([2H])C2=O)c([2H])c1Cl
IUPAC Name 5,5-dideuterio-3-(3,5-dichloro-2,4,6-trideuteriophenyl)-2,4-dioxo-N-propan-2-ylimidazolidine-1-carboxamide
Molecular Weight (g/mol) 335.2
Formula Weight 334.0648
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.