missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ iPrep™ gDNA Forensic Card
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
The iPrep Protocol Card is a flash, 512 KB memory card. Each iPrep Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time.
The following iPrep™ Protocol Cards are available from Invitrogen:
- iPrep™ Forensic Card for use with the iPrep ChargeSwitch Forensic kit and the iPRep™ ChargeSwitch gDNA Buccal kit
-iPrep™ Tissue Card for use with the iPrep ChargeSwitch Tissue Kit
- iPrep(TM) Blood Card for use with the iPrep PureLink Blood kit
The following iPrep™ Protocol Cards are available from Invitrogen:
- iPrep™ Forensic Card for use with the iPrep ChargeSwitch Forensic kit and the iPRep™ ChargeSwitch gDNA Buccal kit
-iPrep™ Tissue Card for use with the iPrep ChargeSwitch Tissue Kit
- iPrep(TM) Blood Card for use with the iPrep PureLink Blood kit
Specifications
Specifications
| Product Type | iPrep™ gDNA Forensic Card |
| Format | Card |
| Description | iPrep™ gDNA Forensic Card, 1 Each, Card Type, 512 KB Memory Card, With Flash, Store at Room Temperature, Used With iPrep™ Purification Instrument, Ideal for Automated Nucleic Acid Purification, DNA Extraction |
| For Use With (Equipment) | iPrep™ purification instrument |
| Quantity | 1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction