missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iPrep™ Card: Buffy Coat

Product Code. 10368393
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10368393

Brand: Invitrogen™ IS10015

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

iPrep™ Card: Buffy Coat

The iPrep™ Card: Buffy Coat is a flash memory card, pre-programmed with the purification protocol for the isolation of gDNA from up to 300ul buffy coat. This protocol allows you to get 20-50ug, highly pure gDNA suitable for the most sensitive multiplex PCR applications.

The following iPrep™ Protocol Cards are also available:
- iPrep™ Card: Buffy Coat
- iPrep™ Card: gDNA Blood
- iPrep™ Card: total RNA
- iPrep™ Card: gDNA Tissue
- iPrep™ Card: Forensic
TRUSTED_SUSTAINABILITY

Specifications

Product Type iPrep™ Buffy Coat Card
Format Card
Description iPrep™ Buffy Coat Card, 1 Each, Card Type, 20 to 50μg Yield, With Flash, Store at Room Temperature, Used With iPrep™ Purification Instrument, Ideal for Automated Nucleic Acid Purification, DNA and RNA Purification and Analysis
For Use With (Equipment) iPrep™ purification instrument
Quantity 1 Ea.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.