missing translation for 'onlineSavingsMsg'
Learn More

Ipragliflozin, TRC

Product Code. 30406605
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406605 500 mg
30405929 100 mg
30398923 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406605 Supplier Toronto Research Chemicals Supplier No. TRCI739475500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 761423-87-4
Molecular Formula C21 H21 F O5 S
Molecular Weight (g/mol) 404.45
Synonym D-Glucitol, 1,5-anhydro-1-C-[3-(benzo[b]thien-2-ylmethyl)-4-fluorophenyl]-, (1S)-, (1S)-1,5-Anhydro-1-C-[3-(benzo[b]thien-2-ylmethyl)-4-fluorophenyl]-D-glucitol, ASP 1941, Ipragliflozin, Suglat
IUPAC Name (2S,3R,4R,5S,6R)-2-[3-(1-benzothiophen-2-ylmethyl)-4-fluorophenyl]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2ccc(F)c(Cc3cc4ccccc4s3)c2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H21FO5S/c22-15-6-5-12(21-20(26)19(25)18(24)16(10-23)27-21)7-13(15)9-14-8-11-3-1-2-4-17(11)28-14/h1-8,16,18-21,23-26H,9-10H2/t16-,18-,19+,20-,21+/m1/s1
Formula Weight 404.1094
Chemical Name or Material Ipragliflozin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.