missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ipragliflozin, MedChemExpress
Description
Ipragliflozin (ASP1941) is an orally active and selective SGLT2 inhibitor with IC50s of 7.38 and 1876 nM, 6.73 and 1166 nM, 5.64 and 1380 nM for human SGLT2 and SGLT1, rat SGLT2 and SGLT1, mouse SGLT2 and SGLT1, respectively. Antidiabetic agent.
Specifications
Specifications
| Chemical Name or Material | Ipragliflozin |
| CAS | 761423-87-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H21FO5S |
| Quantity | 50 mg |
| Synonym | ASP1941 |
| Solubility Information | DMSO : ≥ 100 mg/mL (247.25 mM) ∣H2O : ≥ 0.1 mg/mL (0.25 mM) |
| SMILES | FC1=CC=C([C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2)C=C1CC3=CC(C=CC=C4)=C4S3 |
| Molecular Weight (g/mol) | 404.45 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?