missing translation for 'onlineSavingsMsg'
Learn More

Ipragliflozin, MedChemExpress

Product Code. 16451156
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16451156

Brand: Medchem Express HY14894

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ipragliflozin (ASP1941) is an orally active and selective SGLT2 inhibitor with IC50s of 7.38 and 1876 nM, 6.73 and 1166 nM, 5.64 and 1380 nM for human SGLT2 and SGLT1, rat SGLT2 and SGLT1, mouse SGLT2 and SGLT1, respectively. Antidiabetic agent.

Ipragliflozin (ASP1941) is an orally active and selective SGLT2 inhibitor with IC50s of 7.38 and 1876 nM, 6.73 and 1166 nM, 5.64 and 1380 nM for human SGLT2 and SGLT1, rat SGLT2 and SGLT1, mouse SGLT2 and SGLT1, respectively. Antidiabetic agent.

Specifications

Chemical Name or Material Ipragliflozin
CAS 761423-87-4
Color White
Purity Grade Notes Research
Molecular Formula C21H21FO5S
Quantity 200 mg
Synonym ASP1941
Solubility Information DMSO : ≥ 100 mg/mL (247.25 mM) ∣H2O : ≥ 0.1 mg/mL (0.25 mM)
SMILES FC1=CC=C([C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2)C=C1CC3=CC(C=CC=C4)=C4S3
Molecular Weight (g/mol) 404.45
Formula Weight 404.45
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.