missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IPPK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12603-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a kinase that phosphorylates position 2 of inositol-1, 3, 4, 5, 6-pentakisphosphate to form inositol-1, 2, 3, 4, 5, 6-hexakisphosphate (InsP6). InsP6 has a variety of functions, including stimulation of DNA repair, endocytosis, and mRNA export.Specifications
| IPPK | |
| Polyclonal | |
| Unconjugated | |
| IPPK | |
| C9orf12, Ins(1,3,4,5,6)P5 2 kinase, InsP5 2 kinase, INSP5K2, IP5K, IPK1, IPK1 homolog, IPPK, KIAA0699 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 306808, 64768, 75678 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5PXE9, Q6P1C1, Q9H8X2 | |
| IPPK | |
| IPPK Fusion Protein Ag3278 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction