missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IPPK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554243
This item is not returnable.
View return policy
Description
Immunogen sequence: GCLLYKTLQV QMLDLLDIEG LYPLYNRVER YLEEFPEERK TLQIDGPYDE AFYQKLLDLS TEDDGTVAFA LTKVQQYR Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.
The protein encoded by this gene is a kinase that phosphorylates position 2 of inositol-1, 3, 4, 5, 6-pentakisphosphate to form inositol-1, 2, 3, 4, 5, 6-hexakisphosphate (InsP6). InsP6 has a variety of functions, including stimulation of DNA repair, endocytosis, and mRNA export.
Specifications
| IPPK | |
| Polyclonal | |
| Unconjugated | |
| IPPK | |
| 1,3,4,5,6-pentakisphosphate 2-kinase; 1810043M15Rik; bA476B13.1; bA476B13.1 (novel protein); C9orf12; FLJ13163; hypothetical protein LOC541511; inositol 1,3,4,5,6-pentakisphosphate 2-kinase; inositol 1,3,4,5,6-pentakisphosphate 2-kinase, like; inositol polyphosphate kinase 1; inositol-1,3,4,5,6-pentakisphosphate 2-kinase; Inositol-pentakisphosphate 2-kinase; ins(1,3,4,5,6)P5 2-kinase; InsP5 2-kinase; INSP5K2; InsP6; IP5K; IPK1; IPK1 homolog; ipk1l; ippk; KIAA0699; RGD1311271; rIpk1; si:dkey-42i9.3; zgc:110769 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 306808, 64768, 75678 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5PXE9, Q6P1C1, Q9H8X2 | |
| IPPK | |
| Recombinant protein corresponding to Human IPPK. Recombinant protein control fragment (Product #RP-92947). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction