Learn More
IPPK Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand:  Novus Biologicals H00064768-B01P
Product Code. 18483379
  • £473.00 / 50µg
Description
IPPK Polyclonal antibody specifically detects IPPK in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IPPK
Polyclonal
Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
NP_073592.1
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS (pH 7.4)
bA476B13.1, bA476B13.1 (novel protein), C9orf12, chromosome 9 open reading frame 12, EC 2.7.1.158, FLJ13163, inositol 13,45,6-pentakisphosphate 2-kinase, Inositol-13,45,6-pentakisphosphate 2-kinase, inositol-pentakisphosphate 2-kinase, Ins(13,45,6)P5 2-kinase, InsP5 2-kinase, INSP5K2, IP5K, IPK1, IPK1 homolog, KIAA0699
IPPK (NP_073592.1, 1 a.a. - 491 a.a.) full-length human protein. MEEGKMDENEWGYHGEGNKSLVVAHAQRCVVLRFLKFPPNRKKTSEEIFQHLQNIVDFGKNVMKEFLGENYVHYGEVVQLPLEFVKQLCLKIQSERPESRCDKDLDTLSGYAMCLPNLTRLQTYRFAEHRPILCVEIKPKCGFIPFSSDVTHEMKHKVCRYCMHQHLKVATGKWKQISKYCPLDLYSGNKQRMHFALKSLLQEAQNNLKIFKNGELIYGCKDARSPVADWSELAHHLKPFFFPSNGLASGPHCTRAVIRELVHVITRVLLSGSDKGRAGTLSPGLGPQGPRVCEASPFSRSLRCQGKNTPERSGLPKGCLLYKTLQVQMLDLLDIEGLYPLYNRVERYLEEFPEERKTLQIDGPYDEAFYQKLLDLSTEDDGTVAFALTKVQQYRVAMTAKDCSIMIALSPCLQDASSDQRPVVPSSRSRFAFSVSVLDLDLKPYESIPHQYKLDGKIVNYYSKTVRAKDNAVMSTRFKESEDCTLVLHKV
0.05 mg
Protein Kinase
64768
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.