missing translation for 'onlineSavingsMsg'
Learn More

IPP Antibody (4C2), Novus Biologicals™

Product Code. 18349788 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349788 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349788 Supplier Novus Biologicals Supplier No. H00003652M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IPP Monoclonal antibody specifically detects IPP in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IPP
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4C2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005888.1
Gene Alias actin-binding protein IPP, intracisternal A particle-promoted polypeptideKLHL27Kelch-like protein 27
Host Species Mouse
Immunogen IPP (NP_005888.1, 105 a.a. ~ 204 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NNVQELIIAADMLQLTEVVHLCCEFLKGQIDPLNCIGIFQFSEQIACHDLLEFSENYIHVHFLEVHSGEEFLALTKDQLIKILRSEELSIEDEYQVFLAA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3652
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.