Learn More
IPO7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28289-1-AP-20UL
This item is not returnable.
View return policy
Description
The IPO7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IPO7. This antibody recognizes human antigen. The IPO7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| IPO7 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 10527 | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 467 μg/mL | |
| Rabbit | |
| Primary | |
| Human |