missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IPO13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11696-2-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the importin-beta family of nuclear transport proteins. The encoded protein mediates the import of specific cargo proteins from the cytoplasm to the nucleus and is dependent on the Ras-related nuclear protein-GTPase system. The encoded protein is also involved in nuclear export of the eukaryotic translation initiation factor 1A.Specifications
| IPO13 | |
| Polyclonal | |
| Unconjugated | |
| IPO13 | |
| IMP13, importin 13, IPO13, KAP13, Karyopherin 13, KIAA0724, LGL2, Ran binding protein 13, RANBP13 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116458, 230673, 9670 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O94829, Q8K0C1, Q9JM04 | |
| IPO13 | |
| IPO13 Fusion Protein Ag2317 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction