Learn More
Invitrogen™ IPMK Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101210
Product Code. 16697684
  • £452.00 / 100µL
Description
Antibody detects endogenous levels of total IPMK.
This gene encodes a member of the inositol phosphokinase family. The encoded protein has 3-kinase, 5-kinase and 6-kinase activities on phosphorylated inositol substrates. The encoded protein plays an important role in the biosynthesis of inositol 1, 3, 4, 5, 6-pentakisphosphate, and has a preferred 5-kinase activity. This gene may play a role in nuclear mRNA export. Pseudogenes of this gene are located on the long arm of chromosome 13 and the short arm of chromosome 19.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IPMK
Polyclonal
Unconjugated
IPMK
2410017C19Rik; AA408208; Impk; Inositol 1,3,4,6-tetrakisphosphate 5-kinase; inositol polyphosphate kinase 2; inositol polyphosphate multikinase; Ipk2; Ipmk; LOW QUALITY PROTEIN: inositol polyphosphate multikinase
Rabbit
Affinity Chromatography
RUO
171458, 253430, 69718
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q7TT16, Q8NFU5, Q99NI4
IPMK
A synthesized peptide derived from human IPMK(Accession Q8NFU5), corresponding to amino acid residues S315-V365.
100 μL
Primary
Human, Mouse, Rat