missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ipilimumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
£397.00 - £1002.00
Specifications
| Antigen | Ipilimumab |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30283360
![]() |
Invitrogen™
MA558287 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30282395
|
Invitrogen™
MA558288 |
1 mg |
£1002.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ipilimumab (Yervoy) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with unresectable or metastatic melanoma. Ipilimumab works to activate the immune system by targeting CTLA-4 (Cytotoxic T-lymphocyte protein 4), a negative regulator of cytotoxic T-lymphocyte activity. Ipilimumab binds to CTLA-4, blocking the inhibitory signal, which allows the cytotoxic T-lymphocytes to destroy the cancer cells.Specifications
| Ipilimumab | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| BMS-734016; MDX-010; MDX-CTLA-4; Yervoy | |
| Antibody |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title