missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IPG-4 AM
Click to view available options
Quantity:
1 Pack (10 x 50 μg)
Unit Size:
500µg
Description
IPG-4 AM is a membrane-permeable fluorescent potassium ion (K+) indicator. It binds to potassium (Kd = 7 mM). Excitation and emission maxima (λ) are 525 and 545 nm, respectively. It has 100:1 selectivity for K+ over Na+ ions. It can be used to detect in real time changes in intracellular K+. Also used to monitor extracellular K+ changes in vivo. It is recommended to prepare stock solutions in DMSO.
Specifications
Specifications
| Chemical Name or Material | Acetyloxymethyl 3-[3-(acetyloxymethoxy)-7-[3-(acetyloxymethoxy)-3-oxopropyl]-4,5-dichloro-9-[4-(5,17-dimethyl-8,14,24,27,32,35-hexaoxa-1,11,21-triazatetracyclo[19.8.8.02,7.015,20]heptatriaconta-2(7),3,5,15(20),16,18-hexaen-11-yl)-3-(2-methoxyethoxy)phenyl |
| Quantity | 1 Pack (10 x 50 μg) |
| Target | Ion Indicators |
| Molecular Formula | C67H79Cl2N3O21 |
| Purity | 90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction