missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IPCEF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542835
This item is not returnable.
View return policy
Description
Peptide sequence: IHKALENSFV TSESGFLNSL SSDDTSSLSS NHDHLTVPDK PAGSKIMDKE Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 93%.
Enhances the promotion of guanine-nucleotide exchange by PSCD2 on ARF6 in a concentration-dependent manner. Target protein binds guanine nucleotide exchange factor cytohesin 2 and increases cytohesin 2 mediated formation of ADP-ribosylation factor GTP-ase.
Specifications
| IPCEF1 | |
| Polyclonal | |
| Unconjugated | |
| IPCEF1 | |
| A130090K04Rik; interaction protein for cytohesin exchange factors 1; interaction protein for cytohesin exchange factors 1 Interaction protein for cytohesin exchange factors 1; interactor protein for cytohesin exchange factors 1; Ipcef1; KIAA0403; phosphoinositide binding protein PIP3-E; phosphoinositide-binding protein PIP3-E; PIP3-E; RP3-402L9.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26034 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WWN9 | |
| IPCEF1 | |
| Synthetic peptide directed towards the middle region of human PIP3-E. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction