missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IPCEF1 Antibody (OTI3G2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71781AF488
This item is not returnable.
View return policy
Description
IPCEF1 Monoclonal specifically detects IPCEF1 in Human samples. It is validated for Western Blot.
Specifications
| IPCEF1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IPCEF1 | |
| Full length recombinant protein of human IPCEF1 (NP_056368) produced in HEK293T cells. | |
| 0.1 mL | |
| 26034.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI3G2 | |
| Western Blot | |
| interaction protein for cytohesin exchange factors 1, interactor protein for cytohesin exchange factors 1, KIAA0403interaction protein for cytohesin exchange factors 1 Interaction protein forcytohesin exchange factors 1, phosphoinositide binding protein PIP3-E, Phosphoinositide-binding protein PIP3-E, PIP3-E, RP3-402L9.2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction