missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IP7e

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
IP7e is a potent Nurr1 activator (EC50 = 3.9 nM). Delays onset and reduces severity of symptoms in mice with experimental autoimmune encephalomyelitis (EAE). Also suppresses NF-κB signaling. Brain penetrant and orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 6-[4-[(2-Methoxyethoxy)methyl]phenyl]-5-methyl-3-phenyl-isoxazolo[4,5-c]pyridin-4(5H)-one |
| CAS | 500164-74-9 |
| Quantity | 10 mg |
| Target | NFkB/IkB Inhibitors |
| Molecular Formula | C23H22N2O4 |
| Formula Weight | 390.43 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering