Learn More
Invitrogen™ IP6K1 Recombinant Rabbit Monoclonal Antibody (9P5R5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551386
Description
Immunogen sequence: MCVCQTMEVG QYGKNASRAG DRGVLLEPFI HQVGGHSSMM RYDDHTVCKP LISREQRFYE SLPPEMKEFT PEYKGVVSVC FEGDSDGYIN LVAYPYVESE TVEQDDTTER EQPRRKHSRR SLHRSGSGSD HKEEKASLSL ETSESSQEAK SPKVELHSHS EVPFQMLDGN SGLSSEKISH NPWSLRCHKQ QLSRMRSESK DRKLYKFLLL ENVVHHFKYP.
This gene encodes a member of the inositol phosphokinase family. The encoded protein may be responsible for the conversion of inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). It may also convert 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4. Alternatively spliced transcript variants have been described.
Specifications
| IP6K1 | |
| Recombinant Monoclonal | |
| 0.233 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q6PD10, Q92551, Q9ESM0 | |
| IP6K1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human IP6K1 (NP_695005.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 9P5R5 | |
| Unconjugated | |
| IP6K1 | |
| 1200016D08Rik; ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; AU022776; IHPK1; inositol hexakisphosphate kinase 1; inositol hexakisphosphate kinase 6; inositol hexaphosphate kinase 1; InsP6; InsP6 kinase 1; InsP6k1; IP6K1; Itpk6; KIAA0263; Pi uptake stimulator; PiUS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27399, 50560, 9807 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.