missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IP6K1 Recombinant Rabbit Monoclonal Antibody (3L6N2)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£147.00 - £363.00

Specifications

Antigen IP6K1
Clone 3L6N2
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30282979
View Documents
Invitrogen™
MA558035
20 μL
£147.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284569
GREENER_CHOICE
View Documents
Invitrogen™
MA558036
100 μL
£363.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: MCVCQTMEVG QYGKNASRAG DRGVLLEPFI HQVGGHSSMM RYDDHTVCKP LISREQRFYE SLPPEMKEFT PEYKGVVSVC FEGDSDGYIN LVAYPYVESE TVEQDDTTER EQPRRKHSRR SLHRSGSGSD HKEEKASLSL ETSESSQEAK SPKVELHSHS EVPFQMLDGN SGLSSEKISH NPWSLRCHKQ QLSRMRSESK DRKLYKFLLL ENVVHHFKYP.

This gene encodes a member of the inositol phosphokinase family. The encoded protein may be responsible for the conversion of inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). It may also convert 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4. Alternatively spliced transcript variants have been described.
Specifications

Specifications

IP6K1
1 mg/mL
ELISA, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q6PD10, Q92551, Q9ESM0
27399, 50560, 9807
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human IP6K1 (NP_695005.1).
Antibody
3L6N2
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
IP6K1
1200016D08Rik; ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; AU022776; IHPK1; inositol hexakisphosphate kinase 1; inositol hexakisphosphate kinase 6; inositol hexaphosphate kinase 1; InsP6; InsP6 kinase 1; InsP6k1; IP6K1; Itpk6; KIAA0263; Pi uptake stimulator; PiUS
IP6K1
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.