missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IP6K1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588426

Product Code. 16566671

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MCVCQTMEVG QYGKNASRAG DRGVLLEPFI HQVGGHSSMM RYDDHTVCKP LISREQRFYE SLPPEMKEFT PEYKGVVSVC FEGDSDGYIN LVAYPYVESE TVEQDDTTER EQPRRKHSRR SLHRSGSGSD HKEEKASLSL ETSESSQEAK SPKVELHSHS EVPFQMLDGN SGLSSEKISH NPWSLRCHKQ QLSRMRSESK DRKLYKFLLL ENVVHHFKYP; Positive Samples: HeLa, 293T, HepG2, Mouse brain, Rat brain, Rat testis; Cellular Location: Cytoplasm, Nucleus.

This gene encodes a member of the inositol phosphokinase family. The encoded protein may be responsible for the conversion of inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). It may also convert 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IP6K1
Polyclonal
Unconjugated
IP6K1
1200016D08Rik; ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; AU022776; IHPK1; inositol hexakisphosphate kinase 1; inositol hexakisphosphate kinase 6; inositol hexaphosphate kinase 1; InsP6; InsP6 kinase 1; InsP6k1; IP6K1; Itpk6; KIAA0263; Pi uptake stimulator; PiUS
Rabbit
Affinity Chromatography
RUO
27399, 50560, 9807
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.51 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q6PD10, Q92551, Q9ESM0
IP6K1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human IP6K1 (NP_6950051).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.