missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP6K1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00228-100ul
This item is not returnable.
View return policy
Description
IP6K1 Polyclonal antibody specifically detects IP6K1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| IP6K1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, EC 2.7.4.21, inositol hexakisphosphate kinase 1, Inositol hexaphosphate kinase 1KIAA0263IHPK1PiUS, InsP6 kinase 1, MGC9925, Pi uptake stimulator | |
| Produced in rabbits immunized with purified, recombinant Human IP6K1 (Uniprot#: Q92551-1; Met1-Gln441) | |
| 100 μg | |
| Neuroscience, Neurotransmission, Signal Transduction | |
| 9807 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction