missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP6K1 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18438JF585
This item is not returnable.
View return policy
Description
IP6K1 Polyclonal antibody specifically detects IP6K1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| IP6K1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 585 | |
| ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, EC 2.7.4.21, inositol hexakisphosphate kinase 1, Inositol hexaphosphate kinase 1KIAA0263IHPK1PiUS, InsP6 kinase 1, MGC9925, Pi uptake stimulator | |
| Recognizes region between residues Between Between 75 and 125.. | |
| 0.1 mL | |
| Neuroscience, Neurotransmission, Signal Transduction | |
| 9807 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction