missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP3R3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33409-20ul
This item is not returnable.
View return policy
Description
IP3R3 Monoclonal antibody specifically detects IP3R3 in Human samples. It is validated for ELISA,Western Blot
Specifications
| IP3R3 | |
| Monoclonal | |
| Western Blot 1:2000 - 1:6000, ELISA Recommended starting concentration is 1 μg/mL | |
| FLJ36205, inositol 14,5-triphosphate receptor, type 3, inositol 14,5-trisphosphate receptor type 3, InsP3R3, IP3 receptor, IP3 receptor isoform 3, IP3R, IP3R 3, IP3R3, Type 3 inositol 14,5-trisphosphate receptor, Type 3 InsP3 receptor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1630-1870 of human IP3R3 (NP_002215.2).,, Sequence:, EGSEAYQRCESGGFLSKLIQHTKDLMESEEKLCIKVLRTLQQMLLKKTKYGDRGNQLRKMLLQNYLQNRKSTSRGDLPDPIGTGLDPDWSAIAATQCRLDKEGATKLVCDLITSTKNEKIFQESIGLAIHLLDGGNTEIQKSFHNLMMSDKKSERFFKVLHDRMKRAQQETKSTVAVNMNDLGSQPHEDREPVDPTTKGRVASFSIPGSSSRYSLGPSLRRGHEVSERVQSSEMGTSVLIM | |
| 20 μL | |
| B Cell Development and Differentiation Markers, Immunology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 3710 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction