missing translation for 'onlineSavingsMsg'
Learn More

IP3R3 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18410234 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410234 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410234 Supplier Novus Biologicals Supplier No. NBP121400DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IP3R3 Polyclonal antibody specifically detects IP3R3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP3R3
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ36205, inositol 14,5-triphosphate receptor, type 3, inositol 14,5-trisphosphate receptor type 3, InsP3R3, IP3 receptor, IP3 receptor isoform 3, IP3R, IP3R 3, IP3R3, Type 3 inositol 14,5-trisphosphate receptor, Type 3 InsP3 receptor
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1755 and 1825 of human inositol 1,4,5-triphosphate receptor, type 3 using the numbering given in entry NP_002215.2 (GeneID 3710).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3710
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.