missing translation for 'onlineSavingsMsg'
Learn More

IP3R1 Rabbit anti-Mouse, Rat, Clone: 4Z1B6, Novus Biologicals™

Product Code. 18354815 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354815 20 μg 20µL
18389624 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18354815 Supplier Novus Biologicals Supplier No. NBP31641420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IP3R1 Monoclonal antibody specifically detects IP3R1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP3R1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4Z1B6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DKFZp313E1334, DKFZp313N1434, inositol 14,5-triphosphate receptor, type 1, inositol 14,5-trisphosphate receptor type 1, INSP3R1, IP3 receptor, IP3 receptor isoform 1, IP3R, IP3R 1, IP3R1, SCA15, SCA16, spinocerebellar ataxia 15, spinocerebellar ataxia 16, Type 1 inositol 14,5-trisphosphate receptor, Type 1 InsP3 receptor
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 2200-2300 of human IP3R1 (Q14643).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3708
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.