missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP3KC Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-36962
This item is not returnable.
View return policy
Description
IP3KC Polyclonal specifically detects IP3KC in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| IP3KC | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| inositol 14,5-trisphosphate 3-kinase CIP3K C, inositol-trisphosphate 3-kinase C, InsP 3 kinase C, InsP 3-kinase C, IP3 3-kinase C, IP3-3KC, IP3KCEC 2.7.1.127 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 80271 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, Peptide ELISA Detection limit 1:128000 | |
| NP_079470.1 | |
| ITPKC | |
| Peptide with sequence C-ETERPKQKTEPDRSS corresponding to internal region according to NP_079470.1. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction