missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP 336, Sulfur Set D in Light Oil (20 cSt): S @ 20000, 30000, 40000, 50000 μg/g, VHG™
Click to view available options
Quantity:
4 x 100 mL
Unit Size:
400mL
Description
IP 336, Sulfur Set D in Light Oil (20 cSt): S @ 20000, 30000, 40000, 50000 μg/g
Specifications
Specifications
| Chemical Name or Material | Sulfur |
| CAS | 7704-34-9 |
| Methods | XRF/UVF |
| Analyte or Component Names | Sulfur |
| Shipping Condition | Room Temperature |
| Molecular Formula | S |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/S |
| SMILES | [S] |
| IUPAC Name | sulfur |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction