missing translation for 'onlineSavingsMsg'
Learn More

Ioxitalamic Acid-d4, TRC

Product Code. 30399938
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399938 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30399938 Supplier Toronto Research Chemicals Supplier No. TRCI73810210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ioxitalamic Acid-d4
CAS 1185156-82-4
Molecular Formula C12 H7 D4 I3 N2 O5
Quantity 10 mg
InChI Formula InChI=1S/C12H11I3N2O5/c1-4(19)17-10-8(14)5(11(20)16-2-3-18)7(13)6(9(10)15)12(21)22/h18H,2-3H2,1H3,(H,16,20)(H,17,19)(H,21,22)/i2D2,3D2
SMILES [2H]C([2H])(O)C([2H])([2H])NC(=O)c1c(I)c(NC(=O)C)c(I)c(C(=O)O)c1I
IUPAC Name 3-acetamido-2,4,6-triiodo-5-[(1,1,2,2-tetradeuterio-2-hydroxy-ethyl)carbamoyl]benzoic acid
Molecular Weight (g/mol) 647.96
Formula Weight 647.8053
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.