missing translation for 'onlineSavingsMsg'
Learn More

Ioversol, MedChemExpress

Product Code. 16488698
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488698 10 mg 10mg
16434597 50 mg 50mg
16428756 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488698 Supplier Medchem Express Supplier No. HYB1410
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ioversol (MP-328) is a nonionic iodinated contrast medium (CM) that is used during a CT scan or x-ray in animal experiment. Ioversol does not damage the blood-brain barrier (BBB) in animal.

Ioversol (MP-328) is a nonionic iodinated contrast medium (CM) that is used during a CT scan or x-ray in animal experiment. Ioversol does not damage the blood-brain barrier (BBB) in animal.

Specifications

Chemical Name or Material Ioversol
CAS 87771-40-2
Color White
Purity Grade Notes Research
Molecular Formula C18H24I3N3O9
Quantity 10 mg
Synonym MP-328
Solubility Information DMSO : ≥ 100 mg/mL (123.90 mM)
SMILES O=C(C1=C(I)C(N(C(CO)=O)CCO)=C(I)C(C(NCC(O)CO)=O)=C1I)NCC(O)CO
Molecular Weight (g/mol) 807.11
Formula Weight 807.11
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.