missing translation for 'onlineSavingsMsg'
Learn More

Ioticasone Furoate, TRC

Product Code. 30396746
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396746 2.5 mg 2.5mg
30390701 25 mg 25mg
30396745 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396746 Supplier Toronto Research Chemicals Supplier No. TRCI7363202.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ioticasone Furoate
Analyte or Component Names Ioticasone Furoate
Shipping Condition Room Temperature
Molecular Formula C27 H29 F2 I O6 S
Synonym 6α,9-Difluoro-11β-hydroxy-16α-methyl-17-((iodosulfanyl)carbonyl)-3-oxo-androsta-1,4-diene-17a-yl Furoate
InChI Formula InChI=1S/C27H29F2IO6S/c1-14-9-16-17-11-19(28)18-10-15(31)6-7-24(18,2)26(17,29)21(32)12-25(16,3)27(14,23(34)37-13-30)36-22(33)20-5-4-8-35-20/h4-8,10,14,16-17,19,21,32H,9,11-13H2,1-3H3/t14-,16+,17+,19+,21+,24+,25+,26+,27+/m1/s1
Solution Type Neat
SMILES C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)SCI
IUPAC Name [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11-hydroxy-17-(iodomethylsulfanylcarbonyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 646.482
Quantity 2.5 mg
Formula Weight 646.07
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.