missing translation for 'onlineSavingsMsg'
Learn More

Iothalamic Acid, TRC

Product Code. 30135221
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135221 50 mg 50mg
30004095 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135221 Supplier Toronto Research Chemicals Supplier No. TRCI73620050MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2276-90-6
Molecular Formula C11 H9 I3 N2 O4
Molecular Weight (g/mol) 613.91
Synonym Benzoic acid, 3-(acetylamino)-2,4,6-triiodo-5-[(methylamino)carbonyl]-, Isophthalamic acid, 5-acetamido-2,4,6-triiodo-N-methyl- (7CI,8CI), 3-(Acetylamino)-2,4,6-triiodo-5-[(methylamino)carbonyl]benzoic acid, 5-Acetamido-2,4,6-triiodo-N-methylisophthalamic acid, 5-Acetylamino-2,4,6-triiodo-N-methyl-3-isophthalamic acid, Cysto Conray II, Iotalamic acid, Iothalamic acid, MI 216, Methalamic acid
IUPAC Name 3-acetamido-2,4,6-triiodo-5-(methylcarbamoyl)benzoic acid
SMILES CNC(=O)c1c(I)c(NC(=O)C)c(I)c(C(=O)O)c1I

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H9I3N2O4/c1-3(17)16-9-7(13)4(10(18)15-2)6(12)5(8(9)14)11(19)20/h1-2H3,(H,15,18)(H,16,17)(H,19,20)
Formula Weight 613.7696
Chemical Name or Material Iothalamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.