missing translation for 'onlineSavingsMsg'
Learn More

Iothalamic Acid-d3, TRC

Product Code. 30400464
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400464 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30400464 Supplier Toronto Research Chemicals Supplier No. TRCI7362025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iothalamic Acid-d3
CAS 928623-31-8
Purity Grade Notes HPLC
Molecular Formula C11H6D3I3N2O4
Quantity 5 mg
Synonym Benzoic acid, 3-(acetyl-2,2,2-d3-amino)-2,4,6-triiodo-5-[(methylamino)carbonyl]-, 3-(Acetyl-2,2,2-d3-amino)-2,4,6-triiodo-5-[(methylamino)carbonyl]benzoic acid, Iothalamic acid D3 (acetyl-D3)
InChI Formula InChI=1S/C11H9I3N2O4/c1-3(17)16-9-7(13)4(10(18)15-2)6(12)5(8(9)14)11(19)20/h1-2H3,(H,15,18)(H,16,17)(H,19,20)/i1D3
SMILES [2H]C([2H])([2H])C(=O)Nc1c(I)c(C(=O)O)c(I)c(C(=O)NC)c1I
Molecular Weight (g/mol) 616.93
Percent Purity >95
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.