missing translation for 'onlineSavingsMsg'
Learn More

Iopanoic acid, MedChemExpress

Product Code. 16438796
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438796 Supplier Medchem Express Supplier No. HYB1664
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Iopanoic acid is an inhibitor of 5'-Deiodinase and also an iodinated contrast medium.

Iopanoic acid is an inhibitor of 5'-Deiodinase and also an iodinated contrast medium.

Specifications

Chemical Name or Material Iopanoic acid
CAS 96-83-3
Color White
Purity Grade Notes Research
Molecular Formula C11H12I3NO2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (175.15 mM; Need ultrasonic)
SMILES O=C(O)C(CC)CC1=C(I)C=C(I)C(N)=C1I
Molecular Weight (g/mol) 570.93
Formula Weight 570.93
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.