missing translation for 'onlineSavingsMsg'
Learn More

Iopamidol, MedChemExpress

Product Code. 16472196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472196 500 mg 500mg
16448416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472196 Supplier Medchem Express Supplier No. HYB0684500mg
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Iopamidol is a nonionic, X-Ray iodinated contrast agent (CA) for a wide variety of diagnostic applications. Iopamidol contains amide and hydroxyl functionalities that can be exploited for the generation of the chemical exchange saturation transfer (CEST) contrast.

Iopamidol is a nonionic, X-Ray iodinated contrast agent (CA) for a wide variety of diagnostic applications. Iopamidol contains amide and hydroxyl functionalities that can be exploited for the generation of the chemical exchange saturation transfer (CEST) contrast.

Specifications

Chemical Name or Material Iopamidol
CAS 60166-93-0
Color White
Purity Grade Notes Research
Molecular Formula C17H22I3N3O8
Quantity 500 mg
Synonym B-15000 SQ-13396
Solubility Information H2O : 100 mg/mL (128.69 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (64.34 mM; Need ultrasonic)
SMILES O=C(C1=C(I)C(NC([C@@H](O)C)=O)=C(I)C(C(NC(CO)CO)=O)=C1I)NC(CO)CO
Molecular Weight (g/mol) 777.09
Formula Weight 777.09
Percent Purity 99.74%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.