missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Inline Filter

Product Code. 15365036 Shop All Applied Biosystems Products
Change view
Click to view available options
20 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15365036 20 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15365036 Supplier Applied Biosystems™ Supplier No. 401770

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inline Filter

Peptide Synthesis Accessories for the Models 431A⁄433A

For Research Use Only. Not for use in diagnostics procedures
TRUSTED_SUSTAINABILITY

Specifications

Description Inline Filter, Room Temperature, For Use With 433A Peptide Synthesizer, Store at Room Temperature, For Research Use Only
For Use With (Application) Protein biology, protein sequencing and peptide synthesis, peptide synthesis
Model 431A/433A Model Inline Filter
Quantity 20 Pack(s)
Type Filter

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.