missing translation for 'onlineSavingsMsg'
Learn More

beta-Ionone, 96%, synthetic

Product Code. 10319070 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10319070 500 g Glass bottle 500g
10450341 5 g Glass bottle 5g
10742771 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10319070 Supplier Thermo Scientific Supplier No. 297135000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14901-07-6 | C13H20O | 192.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14901-07-6
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.30
MDL Number MFCD00001549
InChI Key PSQYTAPXSHCGMF-BQYQJAHWSA-N
Synonym beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone
PubChem CID 638014
ChEBI CHEBI:32325
IUPAC Name (E)-4-(2,6,6-trimethylcyclohexen-1-yl)but-3-en-2-one
SMILES CC(=O)\C=C\C1=C(C)CCCC1(C)C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Yellow
Density 0.9440g/mL
Boiling Point 237.0°C to 239.0°C
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5190 to 1.5215
Quantity 500 g
Beilstein 07, 167
Merck Index 10, 4914
Specific Gravity 0.944
Solubility Information Solubility in water: slightly soluble. Other solubilities: solutle in 2 to 3 parts of 70% alcohol, ether,, chloroform and benzene, miscible with absolute alcohol
Formula Weight 192.3
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material β-Ionone, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.