missing translation for 'onlineSavingsMsg'
Learn More

Ionomycin (Calcium Salt), Fisher BioReagents™

Product Code. p-26587
Click to view available options
Quantity:
1 mg
5 mg
Packaging:
Poly Micro Tube
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 10429883

Brand: Fisher BioReagents BP25271

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ionomycin (Calcium Salt)

  •  Suitable for use in electrophoretic separations, specifically polyacrylamide gel electrophoresis (PAGE)
  • This product is a 40% (w/v) solution of high quality acrylamide
  • Free of DNase, RNase, and protease
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56092-82-1
Molecular Formula C41H70CaO9
Molecular Weight (g/mol) 747.08
MDL Number MFCD00083634
InChI Key RULWQOBWZOQCOY-SQNLDGQHSA-M
PubChem CID 71311740
IUPAC Name calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate
SMILES [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O

Specifications

Type Ionomycin Calcium Salt
Physical Form Solid
Melting Point 205°C to 206°C
Packaging Poly Micro Tube
Color White
Quantity 1 mg
Assay Percent Range ≥98.0 %
Formula Weight 747.1
Percent Purity 98.0% min. (by NP TLC), 99.0% min. (by RP HPLC)
Chemical Name or Material Ionomycin, Calcium Salt, From Streptomyces conglobatus
compliance-icons
Product Identifier
  • Ionomycin, Calcium Salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.