missing translation for 'onlineSavingsMsg'
Learn More

Ionomycin, calcium salt, >98%, MP Biomedicals™

Product Code. 11423272 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423272 5 mg 5mg
11413272 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423272 Supplier MP Biomedicals Supplier No. 0215507005
  • ONLINE EXCLUSIVE PRICE
    £546.00
    £511.35 / 5mg
    Save £34.65
    6% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56092-82-1 | Formula: C41H70CaO9 | Weight: 747.08 g/mol

Chemical Identifiers

CAS 56092-82-1
Molecular Weight (g/mol) 747.08
MDL Number MFCD00083634
InChI Key RULWQOBWZOQCOY-SQNLDGQHSA-M
PubChem CID 71311740
IUPAC Name calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate
SMILES [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O

Specifications

Type Ionomycin Calcium Salt
Physical Form Crystalline Powder
Color White
Quantity 5 mg
Assay Percent Range 1
Molecular Formula C41H70CaO9
Formula Weight 747.1
Percent Purity >98%
Grade Not Available
Chemical Name or Material Ionomycin, calcium salt, For HPLC analysis
compliance-icons
Product Identifier
  • Ionomycin Calcium Salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ionomycin Calcium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.